Learn More
Zeiss™ Axio Vert.A1 Inverted Microscope for Phase Contrast
A compact inverted microscope ideal for routine research. This system is configured for transmitted light brightfield and phase contrast.
Brand: Zeiss™ 491237-0003-000
This item is not returnable.
View return policy
Description
- The Axio Vert.A1 is an inverted microscope ideal for advanced routine research and is the only system in its class with such a large range of features but still compact enough to sit directly beside your incubator. The system offers the ultimate flexibility and all standard contrast techniques including brightfield, phase contrast, PlasDIC, iHMC, DIC and epi-fluorescence are available.
- The system has been designed with ergonomics and ease of use in mind. Whether you are sitting or standing, simply use intermediate pieces to work comfortably in an upright position. The ergo tube lets you tilt the eyepieces as much as you need. You can always view your samples easily. Thanks to ergonomic design of all operating parts, your routine tasks and workflows will be easier.
- The Axio Vert.A1 offers either Halaogen or LED illumination for transmitted light. There are then two options for epi-fluorescence - either the Axio Vert.A1 FL-LED with inbuilt LEDs or the Axio Vert.A1 Fl with the ability to use an external light source for fluorescence to give more flexibility.
- The accessories available with the Axio Vert.A1 are vast and these include multiple binocular tubes, documentation ports and attachments for infinity space. There are a range of options for stages with inserts available for a range of samples.
Specifications
LD Condenser 0.4 | |
Phase Contrast |