Learn More
Corning™ AxyGem™ Crystallography Plate
With 96 buffer wells and an adjacent protein well
AxyGem Plate is an SBS standard 96-well sitting-drop, high-throughput protein crystallization plate with 96 buffer wells (maximum volume of 250μL) and one adjacent protein well (maximum volume of 4.3μL)
Brand: Corning™ CP-AXYGEM-96-50
584.47 GBP valid until 2024-12-31
Use promo code "23272" to get your promotional price.
Code : 68
Additional Details : Weight : 3.63000kg
Description
An SBS standard 96-well sitting-drop, high-throughput protein crystallization plate.
- Made of optically clear polystyrene to enable precise crystal visualization
- The buffer well has a novel D-shape geometry located on the right, with elevated round shape protein well located to its left
- Can be filled manually with pipetters or with automated liquid handling workstations
- The plate is sealed with commercially available clear adhesive seals, including Axygen™ UltraClear film
- Compatible with manual pipetters and automated robotic equipments
- Resistant to organic solvents
- Reduced polarization and static
- Plates are stackable
- Bar-coding option
- Round protein well allows easier loop access for crystal manipulation
Specifications
Standard | |
Clear | |
96 | |
96 novel D-shape geometry 250μL buffer wells and an adjacent elevated round shape 4 to 3μL protein well |
8 x 12 | |
Polystyrene | |
Non-sterile |