missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 0.6mL Conical Screw Cap Tubes: Presterilized

Product Code. 12509107 Shop All Axygen Scientific Products
Click to view available options
Color:
Clear
Unit Size:
Pack of 500
This item is not returnable. View return policy

Product Code. 12509107

Brand: Axygen™ SCT45CS

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping of restriction enzymes and other reagents

Axygen microcentrifuge tubes feature a screw cap that provides a tight secure seal. Screw cap tubes are available in a variety of colors with a conical or self-standing bottom. Sterile options are also available.

  • Maximum G-force is 20,000 RCF.
  • RNase-/DNase-free
  • Nonpyrogenic


Tubes, with Cap, Sterile


All sterile screw cap microcentrifuge tubes are pre-assembled with non-looped caps with O-Rings.


Colored-cap versions are also available in sterile format.
TRUSTED_SUSTAINABILITY

Specifications

Cap Style Screw Cap
Certifications/Compliance RNase, DNase, pyrogen-free
Closure Color Clear
Closure Material Polypropylene
Capacity (Metric) 0.6 mL
Color Clear
Material Polypropylene
Format Non-looped
Includes O-ring
No. per Pack 500/Pk.
Packaging 500 Elangated Tubes and caps/Pk.; 8 Packs/Cs.
Sterility Sterile
Graduated No
Closure Type Screw Cap
Bottom Shape Conical
Max. RCF 20,000 x g
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.