Learn More
Axygen™ 0.5 mL Self Standing Screw Cap Tubes
Features O-ring manufactured for resistant to many commonly-used laboratory chemicals
Brand:  Axygen™ SCT-050-SS-V-S
 View more versions of this product
Product Code. 12539027
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
For a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping.
  • Available as presterilized and nonsterile tubes
  • Nonsterile cap tubes have unattached, non-looped caps with O-rings
  • Presterilized cap tubes have non-looped caps attached with O-rings
  • Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
  • Maxymum Recovery™ screw cap tubes available as special order items for assorted tubes
  • Wide range of colored tubes for easy sample identification
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RNase, DNase, pyrogen-free
Violet
0.5 mL
Non-looped
Sterile
Conical
20,000 x g
Polypropylene
Polypropylene
O-ring
Screw Cap