missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 1.5 mL Self-Standing Screw Cap Tubes

Features O-ring manufactured from material that is resistant to many commonly-used laboratory chemicals

Brand:  Axygen™ SCT-150-SS-C-S

249.20 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11380385

This item is not returnable. View return policy

Description

Description

For collection, processing, centrifugation, storage and packaging/shipping.

  • Available as presterilized and nonsterile
  • Nonsterile cap tubes have unattached, non-looped caps with O-rings
  • Pre-sterilized cap tubes have non-looped caps attached with O-rings
  • Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
  • Maxymum Recovery™ screw cap tubes available as special order
  • Caps available in range of colors for easy sample identification

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Screw Cap
20,000 x g
Polypropylene
Clear
Non-looped
500/Pk.
No
Conical
RNase, DNase, pyrogen-free
Clear
1.5 mL
Polypropylene
O-ring
Sterile
Screw Cap
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote