Learn More
Axygen™ 1.5 mL Self-Standing Screw Cap Tubes
Features O-ring manufactured from material that is resistant to many commonly-used laboratory chemicals
Brand:  Axygen™ SCT-150-SS-V
 View more versions of this product
Product Code. 12719558
Description
Description
For collection, processing, centrifugation, storage and packaging/shipping.
  • Available as presterilized and nonsterile
  • Nonsterile cap tubes have unattached, non-looped caps with O-rings
  • Pre-sterilized cap tubes have non-looped caps attached with O-rings
  • Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
  • Maxymum Recovery™ screw cap tubes available as special order
  • Caps available in range of colors for easy sample identification
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RNase, DNase, pyrogen-free
Violet
1.5 mL
Non-looped
Non-sterile
Screw Cap
20,000 x g
Polypropylene
Polypropylene
O-ring
No
Conical