missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 10μL Microvolume Pipette Tips

Product Code. 12619535 Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
0.5 to 10 μL
Unit Size:
Pack of 960

Product Code. 12619535

Brand: Axygen™ T300RS

Only null left
Add to basket
Add to basket

Axygen Universal Tips are designed to fit a wide variety of single and multi-channel pipettes, compatible with most popular brands of pipettes. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.

  • Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free
  • Nonpyrogenic
  • Microvolume
  • Maxymum Recovery technology for ultra-low retention and high yield
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 0.5 to 10 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Certifications/Compliance USP 3127
Clean Claims DNase, Rnase Free, Nonpyrogenic
For Use With (Application) Allows recovery of costly samples with improved precision and accuracy
Format Rack
Tip Style Micro Point
For Use With (Equipment) Gilson Pipetman P10, Gilson Pipetman P2
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.