Learn More
Axygen™ 120μm CyclerSeal Sealing Film for Storage and PCR Applications, Nonsterile
Axygen™ microplate sealing films are designed for applications ranging from PCR and Real-time PCR to ELISA and cell culture. They can be used in a wide range of temperatures, making them suitable for nearly any application.
Brand: Axygen™ PCR-TS-900
Code : 68
Additional Details : Weight : 1.36000kg
Description
- This polypropylene film is the easiest and most cost-effective method for sealing plates
- Exceptional seal across entire plate safeguards samples during transport or storage
- When used with Axygen compression mat, eliminates well to well contamination and cross-over in PCR application
- PCR-TS-900 is suitable for water-bath and other PCR application
Specifications
Polypropylene | |
250 | |
Sealing Film |
Non-sterile | |
50 | |
120 μm |