Learn More
Axygen™ 20 μL Automation Tips for Agilent™/Velocity 11 VPrep™ and Bravo, 96 Tip Format
GREENER_CHOICEBIOPHARMA
For use with Agilent/Velocity11 automation systems
Brand:  Axygen™ VTF-20-R-S
 View more versions of this product
Product Code. 12796759
  • £560.00 / Pack of 4800
Estimated Shipment: 20-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
  • Clear polypropylene
  • RNase-/DNase-free
  • Nonpyrogenic
  • Available in nonsterile or sterile
TRUSTED_SUSTAINABILITY
Specifications
Specifications
20 μL
Sterile
ANSI
Non-pyrogenic, used for stringent specifications under strict process controls
Rack
SBS Rack
Agilent/Velocity VPrep 8LT, Agilent/Velocity VPrep 96LT, Agilent Bravo 96LT
Filtered
Clear
DNase, Rnase Free
w/out Bottom Label
Automation, Round
50 Racks/Cs.
Polyethylene