missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ 20 μL Automation Tips for Agilent™/Velocity 11 VPrep™ and Bravo, 96 Tip Format
For use with Agilent/Velocity11 automation systems
Brand: Axygen™ VTF-20-R-S
This item is not returnable.
View return policy
Description
Tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
- Clear polypropylene
- RNase-/DNase-free
- Nonpyrogenic
- Available in nonsterile or sterile
Specifications
20 μL | |
Sterile | |
ANSI | |
Non-pyrogenic, used for stringent specifications under strict process controls | |
Rack | |
SBS Rack | |
Agilent/Velocity VPrep 8LT, Agilent/Velocity VPrep 96LT, Agilent Bravo 96LT |
Filtered | |
Clear | |
DNase, Rnase Free | |
w/out Bottom Label | |
Automation, Round | |
50 Racks/Cs. | |
Polyethylene |