missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ 200μL Universal Pipette Tips, Beveled, Reference, Maxymum Recovery
Designed to fit a wide variety of single and multi-channel pipettes
£66.15 - £525.00
Specifications
Volume (Metric) | 1 to 200 μL |
---|---|
Color | Clear |
Autoclavable | Autoclavable |
Tip Style | Beveled, Low Retention, Universal |
For Use With (Equipment) | Universal Fit |
Product Code | Brand | Format | Sterility | Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Format | Sterility | Color | Price | Quantity & Availability | |||
12571368
|
Axygen™
TR-222-C-L |
Bulk | Non-sterile | Clear |
£525.00
Pack of 20000 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
12505547
|
Axygen™
TR-222-C-L-R |
Rack | Non-sterile | Clear |
£66.15
Pack of 960 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
12515547
|
Axygen™
TR-222-C-L-R-S |
Rack | Sterile | Clear |
£84.45
Pack of 960 |
In Stock
Log in to see stock. |
|||
12591298
|
Axygen™
TR-222-C-L-STK-S |
Rack | Sterile | Clear |
£298.00
Pack of 4800 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
12786629
|
Axygen™
TR-222-C-L-R-S |
Rack | Sterile | Clear |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
13128053
|
Axygen™
TR-222-C-L-R |
Rack | Non-sterile | Clear |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Optimized tip bevel reduces sample hang-up, improves pipetting accuracy
- Maxymum recovery allows complete retrieval of precious samples, saves costly reagents
- Wide bore tips of orifice size: 0.062 in. (1.52mm) makes it ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
- Graduation marks allow a visual confirmation of pipettor draw volume
- Presterilized tips are e-beam irradiated to prevent product degradation
- Clean-room manufacturing environment certifies products safe from RNase, DNase and pyrogen safe
Specifications
1 to 200 μL | |
Autoclavable | |
Universal Fit |
Clear | |
Beveled, Low Retention, Universal | |
Polypropylene |