missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Maxymum Recovery™ 300 μL Universal Pipetter Tips

Product Code. 12541348 Shop All Axygen Scientific Products
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Bulk
Rack
Unit Size:
Pack of 1000
Pack of 10000
Pack of 4800
Pack of 960
This item is not returnable. View return policy

Product Code. 12541348

Brand: Axygen™ T350CL

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Maxymum Recovery tips fit popular brands of pipettes

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.

  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Fine-Point
  • Maxymum Recovery technology for ultra-low retention and high yield

Order Info

  • Bulk: 1000 tips/pack, 240 packs/case
  • Racked: 96 tips/rack, 10 racks/pack, 5 packs/case

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 to 300 μL
Sterility Non-sterile
Color Clear
Clean Claims DNase, RNase, ATP, and Endotoxin Free
Format Bulk
Tip Style Low Retention, Universal
Packaging 1,000 Tips/Pk., 10 Packs/Cs.
For Use With (Equipment) Universal Fit
Product Line Maxymum Recovery
Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.