Learn More
Axygen™ 384-well Skirted PCR Microplates
Single and double notch 384-well PCR plates
Brand:  Axygen™ PCR-384-R
 View more versions of this product
Product Code. 11310165
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Sealing Options: Axymats, heat sealing, adhesive films and foils.
  • Available in 50μL and 55μL
  • Single and double notch corner
Skirted (50μL)
  • Application/Compatibility: standard thermal cycling
  • Robotic-friendly full skirt design
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.
Skirted (55μL)
  • Application/Compatibility: standard thermal cycling, fully ABI compatible for qPCR and sequencing
  • Robotic-friendly full skirt design
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.
Skirted, RigidPlate (50μL)
  • Robotic-friendly rigid plate with full skirt
  • Application/Compatibility: standard thermal cycling, ABI sequencing, automation applications
  • Packaging Description: 10 plates/Pk., 2 packs/sleeve, 5 sleeves/Cs.
Skirted, elevated wells
  • Application/Compatibility: standard thermal cycling, ABI sequencing, qPCR
  • Raised wells allow 55μL volume
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.
Colors available by special order.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RNase, DNase-free
16 x 24
Full
40 μL
40 μL
Round
Double notch
Standard
Red
Polypropylene
384
Non-sterile
PCR
Red