Learn More
Axygen™ 96-Well PCR Microplates
PCR™ products specifically designed for use within PCR thermal transfer and real time PCR applications.
Brand:  Axygen™ PCR-96-C-S
305.86 GBP valid until 2024-12-31
Use promo code "23272" to get your promotional price.
Code : 68
Additional Details : Weight : 1.50000kg
Product Code. 12749448
  • £334.00 / Pack of 50
Estimated Shipment: 20-11-2024
to see stock.
Add to basket
Description
Description
PCR™ products specifically designed for use within PCR thermal transfer and real time PCR applications. A variety of colors are offered to help organize multi-step PCR processes.
  • 96-well PCR microplates
  • Working volume is 300μL
  • Available in a variety of colors
  • Sold 10 plates/unit; 5 units/Cs.
Specifications
Specifications
RNase, DNase-free
8 x 12
Polypropylene
Non-sterile
Clear
Standard
Clear
96
PCR thermal transfer, RT-PCR applications