missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Automation Tips for Beckman Coulter™ 200μL, Clear, Wide Bore, Filtered, Sterile

Compatible with the original equipment manufacturers' tips

Brand:  Axygen™ FXF-205-WB-R-S

Additional Details : Weight : 9.40000kg

Product Code. 15733997

  • £785.00 / Pack of 4800
Estimated Shipment: 11-12-2024
to see stock.
Add to basket

Description

Description

  • Tip format: 96 tip
  • Tip volume range: 5 μL to 250 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
    - Standard or Axygen™ Maxymum Recovery™ surface
  • Compatible heads: AP-96, MP20, MP200
  • Rack format: ANSI/SLAS
  • Rack material: Polypropylene
Specifications

Specifications

Wide Bore
Filtered
Clear
Optimized for automation
Automation, Wide Bore
50 Racks/Cs.
Polypropylene
200 μL
Sterile
DNase, RNase Free
Rack
SBS Rack
Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Biomek 3000, Beckman Coulter Multimek AP96, Beckman Coulter Multimek SPAN-8, Beckman Coulter Multimek MP200
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers