Learn More
Axygen™ AxyMats™ Sealing Mat for 96 Well PCR Microplates
Minimize evaporation in wide range of temperatures in PCR and storage applications
Brand:  Axygen™ AM-96-PCR-RD
335.86 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Product Code. 12577897
  • £475.00 / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Minimize evaporation in wide range of temperatures in PCR and storage applications
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications. AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved. They can be reused following a bleach wash and ethanol rinse protocol.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
Non-sterile
96
10/Pk.
Round
Silicone
Autoclavable
50/Cs.
Sealing Mat
96-well PCR Microplate