Learn More
Axygen™ Axypet™ Pro 12 Multi-Channel Pipettor
GREENER_CHOICE
Product Code. 16372257 Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
0.5 to 10 μL
20 to 200 μL
30 to 300 μL
5 to 50 μL
Color:
Green
Red
Yellow
Unit Size:
Each
Product Code. 16372257
Brand: Axygen™ AP1210P
Only null left
Add to basket
Add to basket
Corning™ Axygen™ Axypet™ Pro Pipettors are engineered to provide the highest levels of comfort, accuracy, and precision.
Axygen™ Axypet™ Pro Pipettors have been engineered to provide the highest levels of ergonomics, accuracy, and precision. Lightweight construction, contoured shape of the handle, and 4-digit counter were designed to ensure comfortable pipetting. All pipettors feature smooth plunger movement and extremely low pipetting forces to reduce the wrist strain and fatigue (RSI). Colored pushbuttons (included) can help define user or application for sample safety and lower risk of cross contamination.
  • Ergonomic handle design for perfect fit for both right and left hand
  • Convenient one-hand volume setting with auto-lock system prevents accidental volume change during work
  • 4-digit counter for greater precision, perfectly positioned for visibility at all times
  • Single-channel pipettors are compatible with very narrow tubes
  • Fully autoclavable and UV-resistant for sample protection against contaminations
  • The multi-channel shafts retract individually for perfect tip loading and easier tip ejection
  • Easy in-lab calibration helps you quickly adjust your pipettor to various liquid types
  • Individually tested and supplied with a certificate of quality
Learn more from North America
Learn more from Europe, Middle East, and Africa
TRUSTED_SUSTAINABILITY
Specifications
Accuracy ±10.0 to ±2.0
Certifications/Compliance ISO8655
Autoclavable Autoclavable
Color Red
Number of Channels 12
Product Line Axypet Pro
Type Pipette
Volume (Metric) 0.5 to 10 μL
Finger Style Ergonomic
Includes Instruction manual, QC Certificate, In-lab calibration tool
Increments 0.05 μL
Precision ≤2.5 to ≤0.6
Quantity 1
Warranty 3
Show More Show Less
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato