Learn More
Axygen™ Axypet™ Pro Pipettor Starter Kit
Corning™ Axygen™ Axypet™ Pro Pipettors are engineered to provide the highest levels of comfort, accuracy, and precision.
Brand: Axygen™ AP-STR-KIT-P
963.39 GBP valid until 2024-12-31
Use promo code "23272" to get your promotional price.
Additional Details : Weight : 1.18000kg
Description
Axygen™ Axypet™ Pro Starter Kit contains 4 variable volume pipettors in the most popular and universal volumes (0.5-10 μL, 2-20 μL, 20-200 μL, and 100-1000 μL), pipet stand for 4 pipettors, and 3 racks with tips.
- Cat. No. AP-10-P Axygen Axypet Pro Single-channel Pipettor, 0.5 to 10 μL
- Cat. No. AP-200-P Axygen Axypet Pro Single-channel Pipettor, 20 to 200 μL
- Cat. No. AP-1000-P Axygen Axypet Pro Single-channel Pipettor, 100 to 1000 μL
- Cat. No. AP-USD4-P Universal linear stand for four single-channel pipettors
- Cat. No. MR-10XT-R (10 μL) Axygen Multirack pipet tips
- Cat. No. MR-200-R (200 μL) Axygen Multirack pipet tips
- Cat. No. MR-1000XT-R (1000 μL) Axygen
Specifications
ISO8655 | |
Red/Yellow/Blue | |
1 | |
Starter Kit | |
4 single-channel pipettors, Linear stand, Sample tips, Instruction manual, QC Certificate, In-lab calibration tool | |
3 |
Autoclavable | |
Ergonomic | |
Axypet | |
Ergonomic | |
1 Kit |