Learn More
Axygen™ Biomek™ FX/NX Aerosol Barrier Filtered Robotic Tips
Clear polypropylene filter tips; autoclavable.
Brand:  Axygen™ FXF-384-XL-R-S
Code : 68
Additional Details : Weight : 10.90000kg
 View more versions of this product
Product Code. 12776699
  • £5590.00 / Pack of 19200
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
Description
Description
  • Tip volume range: 5 to 250 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • non-pyrogenic
  • Available options:
    • With or without aerosol-resistant filters
    • non-sterile or sterile
    • Standard or Axygen™ Maxymum Recovery™ surface
  • Rack format: ANSI/SLAS
  • Rack material: Polypropylene
Specifications
Specifications
30 μL
Sterile
Green
DNase, RNase and Pyrogen free
Rack, 384-well
10 Racks/Pk, 5 Pks/Cs.
Beckman Biomek
Filtered
Clear
Not Autoclavable
Non-pyrogenic, used for stringent specifications under strict process controls
Automation, Extended Length
Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Multimek AP384, Dynamic Devices Oasis, Dynamic Devices Oasis LM 384-Channel Low Volume, Hamilton
Polypropylene