missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Biomek™ FX/NX Aerosol Barrier Filtered Robotic Tips

Clear polypropylene filter tips; autoclavable.

Brand:  Axygen™ FXF-50-R-S

611.14 GBP valid until 2024-12-31
Use promo code "23272" to get your promotional price.



Code : 68

Additional Details : Weight : 9.53000kg

 View more versions of this product

Product Code. 12796699

  • £672.00 / Pack of 4800
Estimated Shipment: 11-12-2024
to see stock.
Add to basket

Description

Description

  • Tip volume range: 5 to 250 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • non-pyrogenic
  • Available options:
    • With or without aerosol-resistant filters
    • non-sterile or sterile
    • Standard or Axygen™ Maxymum Recovery™ surface
  • Rack format: ANSI/SLAS
  • Rack material: Polypropylene
Specifications

Specifications

50 μL
Sterile
Green
DNase, RNase and Pyrogen free
Rack, 96
10 Racks/Pk, 5 Pks/Cs.
Beckman Biomek
Filtered
Clear
Not Autoclavable
Non-pyrogenic, used for stringent specifications under strict process controls
Automation, Round
Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Biomek 3000, Beckman Coulter Multimek AP96, Beckman Coulter Multimek SPAN-8, Beckman Coulter Multimek MP20, Beckman Coulter Multimek MP200
Polypropylene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers