missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Biomek™ FX/NX Robotic Tips
GREENER_CHOICE

Product Code. 12575727 Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
10 μL
20 μL
30 μL
45 μL
50 μL
250 μL
Sterility:
Non-sterile
Sterile
Tip Style:
Automation, Fine Point
Automation, Round
Automation, Wide Orifice
Unit Size:
Pack of 19200
Pack of 3840
Pack of 4800
Pack of 960
This item is not returnable. View return policy

Product Code. 12575727

Brand: Axygen™ FX384R

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Axygen robotic tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.

The Axygen brand range of products includes a comprehensive line of tips for Beckman Coulter automated liquid handling systems. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. The pipette tips are also available with MAXYMum Recovery, a unique low-retention technology that improves tip performance and maximizes the use of your precious samples.

  • Available in racked, racked sterile and with aerosol barrier (filtered)
  • Compatible with Beckman Biomek FX and Beckman Multimek instruments with an AP96 head
  • Also compatible with Span-8 pod when Liquid Level Sensing function is turned off
  • Unsurpassed straightness to ensure tip enters the well smoothly, every time
  • 384/1536-well compatible to meet your most demanding applications
  • Wide bore tips are ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
  • Fine Point improves accuracy and precision at lower sample volumes
  • Maxymum recovery™ tips reduce sample loss, improves reproducibility and accuracy, minimizes usage of precious samples and reagents

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 30 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Not Autoclavable
Certifications/Compliance ANSI
Clean Claims DNase, RNase and Pyrogen free
For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls
Format Rack, 384-well
Tip Style Automation, Round
Packaging 10 Racks/Pk, 5 Pks/Cs.
For Use With (Equipment) Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Biomek 3000, Beckman Coulter Multimek AP96, Beckman Coulter Multimek SPAN-8, Beckman Coulter Multimek MP20, Beckman Coulter Multimek MP200
Product Line Beckman Biomek
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.