Learn More
Axygen™ Biomek™ FX/NX Robotic Tips
BIOPHARMA
Axygen robotic tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
Brand:  Axygen™ FX-250-R
 View more versions of this product
Product Code. 12732856
  • £311.93 / Pack of 4800
Estimated Shipment: 09-09-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The Axygen brand range of products includes a comprehensive line of tips for Beckman Coulter automated liquid handling systems. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. The pipette tips are also available with MAXYMum Recovery, a unique low-retention technology that improves tip performance and maximizes the use of your precious samples.
  • Available in racked, racked sterile and with aerosol barrier (filtered)
  • Compatible with Beckman Biomek FX and Beckman Multimek instruments with an AP96 head
  • Also compatible with Span-8 pod when Liquid Level Sensing function is turned off
  • Unsurpassed straightness to ensure tip enters the well smoothly, every time
  • 384/1536-well compatible to meet your most demanding applications
  • Wide bore tips are ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
  • Fine Point improves accuracy and precision at lower sample volumes
  • Maxymum recovery™ tips reduce sample loss, improves reproducibility and accuracy, minimizes usage of precious samples and reagents
TRUSTED_SUSTAINABILITY
Specifications
Specifications
250 μL
Non-sterile
Not Autoclavable
DNase, RNase and Pyrogen free
Rack, 96
10 Racks/Pk, 5 Pks/Cs.
Beckman Biomek
Non-filtered
Clear
ANSI
Non-pyrogenic, used for stringent specifications under strict process controls
Automation, Round
Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Biomek 3000, Beckman Coulter Multimek AP96, Beckman Coulter Multimek SPAN-8, Beckman Coulter Multimek MP20, Beckman Coulter Multimek MP200
Polypropylene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number17521017 Certificate TypeCertificate of Compliance Date11/09/2023 Catalog NumberFX-250-R
Lot Number30101024 Certificate TypeCertificate of Compliance Date11/09/2023 Catalog NumberFX-250-R
Lot Number26221027 Certificate TypeCertificate of Compliance Date01/09/2023 Catalog NumberFX-250-R
Lot Number26121043 Certificate TypeCertificate of Compliance Date01/09/2023 Catalog NumberFX-250-R
Lot Number25821008 Certificate TypeCertificate of Compliance Date01/09/2023 Catalog NumberFX-250-R
Special Offers
Special Offers
Product Content Correction
Product Title
Axygen™ Biomek™ FX/NX Robotic Tips > 96-well tips; 250 μL; Non-sterile, Round, Clear