Learn More
Axygen™ Caps for PCR Tubes
8-Strip domed or flat strip caps for Axygen PCR tubes
Brand:  Axygen™ PCR-02CP-C
 View more versions of this product
Product Code. 10557330
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Axygen polypropylene, thin wall PCR tubes with and without caps are designed to fit standard 96-well blocks and are available in an assortment of colors. The consistent and ultra-thin wall allows precise thermal transfer. An even insertion cap ensures a tight fit to reduce evaporation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RNase, DNase, pyrogen-free
Clear
125/Pk.
8-strips PCR tubes and 96-well PCR microplates
Polypropylene
Strip
Clear
0.2 mL
Polypropylene
Non-sterile
Domed
8-Strip Domed PCR Tube Cap