Learn More
Axygen Caps, PCR Tube; Axygen; 0.2mL; Thin wall; Domed 8 strip caps; Green; Fit 8 Strip PCR tubes/96-well PCR microplates; Certified RNase-/DNase-free; Nonpyrogenic; Nonsterile; Virgin polyethylene; 125 Strips/Pk.; 10 Pk./Cs.
Brand: Axygen PCR-02CP-G
Caps, PCR Tube; Axygen; 0.2mL; Thin wall; Domed 8 strip caps; Green; Fit 8 Strip PCR tubes/96-well PCR microplates; Certified RNase-/DNase-free; Nonpyrogenic; Nonsterile; Virgin polyethylene; 125 Strips/Pk.; 10 Pk./Cs.
Code : A0
Additional Details : Weight : 0.01000kg