missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Corbett™ Compatible Filter Tips
For use with Corbett X-tractor gene
Brand: Axygen™ CTF-199-R-S
Code : 68
Additional Details : Weight : 9.03000kg
Description
Axygen™ offers a comprehensive line of tips for Corbett X-tractor. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. Tips are RNase-/DNase-free and nonpyrogenic. Available in sterile and filtered. Clear polypropylene tips are available in 96-well format designed for Corbett X-tractor Gene 8-Channel Head.Specifications
199 μL | |
Sterile | |
Rack | |
SBS Rack | |
Corbett |
Filtered | |
Clear | |
Round | |
Corbett™ with X-Tractor Gene | |
Polyethylene |