Learn More
Axygen™ Disposable V-Bottom Reagent Reservoirs
Designed to accommodate all multichannel pipettor
Brand: Axygen™ RES-2CV-25-S
This item is not returnable.
View return policy
Description
Designed to accommodate all multichannel pipettor
Axygen disposable reservoirs are designed to accommodate all multichannel pipettors. Four corners and a sharp V-bottom shape ensure maximum sample retrieval. All reservoirs are white.
Specifications
ANSI, RNase/DNase-free, non-pyrogenic | |
25 mL | |
Sterile | |
Divided; 200/Cs. |
White | |
Polystyrene | |
Reagent Reservoir | |
Single-use |
Product Content Correction
Product Title
Axygen™ Disposable V-Bottom Reagent Reservoirs > 25 mL; Yes; 200/Cs
Spot an opportunity for improvement?Share a Content Correction