missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Disposable V-Bottom Reagent Reservoirs

Designed to accommodate all multichannel pipettor
Brand: Axygen™ RES-2CV-25-SI
100.55 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Designed to accommodate all multichannel pipettor
Axygen disposable reservoirs are designed to accommodate all multichannel pipettors. Four corners and a sharp V-bottom shape ensure maximum sample retrieval. All reservoirs are white.

Specifications
ANSI, RNase/DNase-free, non-pyrogenic | |
25 mL | |
Sterile | |
100/Cs. | |
Divided; Individually Wrapped; 100/Cs. |
White | |
Polystyrene | |
Reagent Reservoir | |
100/Pk. | |
Single-use |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Axygen™ Disposable V-Bottom Reagent Reservoirs > 25 mL; Yes; 100/Cs
Spot an opportunity for improvement?Share a Content Correction