missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Disposable V-Bottom Reagent Reservoirs
GREENER_CHOICE

Designed to accommodate all multichannel pipettor

Brand:  Axygen™ RES-V-100-S

180.27 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 12547997

  • £214.00 / Pack of 200
Estimated Shipment: 20-08-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Designed to accommodate all multichannel pipettor

Axygen disposable reservoirs are designed to accommodate all multichannel pipettors. Four corners and a sharp V-bottom shape ensure maximum sample retrieval. All reservoirs are white.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ANSI, RNase/DNase-free, non-pyrogenic
100 mL
Sterile
200/Cs.
200/Cs.
White
Polystyrene
Reagent Reservoir
5/Pk.
Single-use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Axygen™ Disposable V-Bottom Reagent Reservoirs > 100 mL; Yes; 200/Cs

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.