missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Disposable V-Bottom Reagent Reservoirs

Designed to accommodate all multichannel pipettor
Brand: Axygen™ RES-V-50
80.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Designed to accommodate all multichannel pipettor
Axygen disposable reservoirs are designed to accommodate all multichannel pipettors. Four corners and a sharp V-bottom shape ensure maximum sample retrieval. All reservoirs are white.

Specifications
ANSI, RNase/DNase-free, non-pyrogenic | |
50 mL | |
Non-sterile | |
100/Cs. | |
100/Cs. |
White | |
Polystyrene | |
Reagent Reservoir | |
100/Pk. | |
Single-use |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Axygen™ Disposable V-Bottom Reagent Reservoirs > 50 mL; No; 100/Cs
Spot an opportunity for improvement?Share a Content Correction