Learn More
Axygen™ EasyPress Sealing Mat and Film Applicator
GREENER_CHOICE
Provides simple and effective application of Axymats and ImpermaMats
Brand:  Axygen™ IT-EP-R
1847.56 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 12683926
  • £2545.00 / Each
Explore more special offers
Description
Description
Axygen™ EASY PRESS is a manual device which provides simple and effective application of Axymats and/or Impermamats to 24, 48, 96 or 384 well microplates, deep well plates, microtiter tube systems and PCR plates.The EASY PRESS can be quickly adjusted to handle any size plate of varying heights (384 PCR to 2mL deep well blocks) using the large thumbwheel.
  • Suitable for deep well plates, PCR microplates in 96 and 384 formats
  • Flexible-can be used with plates of various heights and types
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Sealer
    Axymats, deep well plates, microplates, pcr plates
    1 Ea.
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents Certificates
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    Axygen™ EasyPress Sealing Mat and Film Applicator > Sealer; For AxyMats