Learn More
Axygen™ Gel Documentation System Accessory, Orange Glass
For use with Gel Documentation System (GD-1000)
Brand: Axygen™ GD-OG
This item is not returnable.
View return policy
Description
Ideal for reducing background when cutting out bands with blue lightSpecifications
Gel Documentation System (GD-1000) | |
Orange Glass |