missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Microplate Sealing Film and Tapes
Shop All Axygen Scientific Products

Click to view available options
Material:
Aluminum
Litto or Ethylene or Vinyl
Polyester
Polyolefin
Rayon
Product Type:
AxySeal Sealing Film
Breathable Sealing Film
CyclerSeal Sealing Film
CyclerSeal Sealing Film Roll
Heat Seal Film
Sealing Film
Sealing Film Roll
UltraClear Sealing Film
Dimensions (L x W):
115 x 500 m
127.2 x 79.5 mm
133.3 x 79.2 mm
135.5 x 80 mm
146 x 79.2 mm
146 x 79.6 mm
153 x 82.3 mm
78 x 610 m
Not Available
Sterility:
Non-sterile
Sterile
Unit Size:
Each
Pack of 100
Pack of 1000
Pack of 50
Pack of 500
Description
For applications ranging from PCR and Real-Time PCR to ELISA and cell culture, Axygen films are an easy and cost-effective way to seal plates and enhance automation. Used to seal multi-well microplates.
Aluminum
- Provides exceptional seal on multi-well plates with varied temperatures and reagents
- With compression mats, films and tapes seal during thermocycling
- Ideal for light-sensitive samples and its uniformly applied adhesive provides optimum well sealing
- Pierceable, PCR-AS-200
Specifications
Specifications
| Material | Aluminum |
| Sterility | Non-sterile |
| Length (English) | 4.52 in. |
| Length (Metric) | 115 mm |
| No. per Case | 1/Cs. |
| No. per Pack | 1/Pk. |
| Dimensions (L x W) | 115 x 500 m |
| Width (English) | 196 in. |
| Width (Metric) | 500 m |
| For Use With (Application) | Designed for applications ranging from PCR and real-time PCR to storage, ideal for light-sensitive samples |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction