missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Microplate Sealing Film and Tapes
Shop All Axygen Scientific Products
Click to view available options
Material:
Aluminum
Litto or Ethylene or Vinyl
Polyester
Polyolefin
Rayon
Product Type:
AxySeal Sealing Film
Breathable Sealing Film
CyclerSeal Sealing Film
CyclerSeal Sealing Film Roll
Heat Seal Film
Sealing Film
Sealing Film Roll
UltraClear Sealing Film
Dimensions (L x W):
115 x 500 m
127.2 x 79.5 mm
133.3 x 79.2 mm
135.5 x 80 mm
146 x 79.2 mm
146 x 79.6 mm
153 x 82.3 mm
78 x 610 m
Not Available
Sterility:
Non-sterile
Sterile
Unit Size:
Each
Pack of 100
Pack of 1000
Pack of 50
Pack of 500
Description
For applications ranging from PCR and Real-Time PCR to ELISA and cell culture, Axygen films are an easy and cost-effective way to seal plates and enhance automation. Used to seal multi-well microplates. Sealing film temperature ranges from -40°C to +104°C.
Breathable
- Allow effective gas exchange for cellular and bacterial cultivation — while preventing contamination
- Seal polypropylene and polystyrene culture plates, 96- and 384-well plates including other assay plates
- Provides exceptional seal on multi-well plates with varied temperatures and reagents
- With compression mats, films and tapes seal during thermocycling
- Ideal for light-sensitive samples and its uniformly applied adhesive provides optimum well sealing
- Pierceable, PCR-AS-200
- Exceptional seal across entire plate safeguards samples during transport or storage
- When used with Axygen's compression mat, this polyester film eliminates well-to-well contamination and cross-over in PCR applications
- Polyester-based with uniformly and consistently applied acrylic adhesive to eliminate the edge effect in sensitive ELISA assays
- Suitable for sealing tissue culture plates, short-term storage and incubation, transport and the containment of biohazardous solutions
- Available in both nonsterile and sterile packaging
Specifications
Specifications
| Material | Polyester |
| Sterility | Non-sterile |
| Length (English) | 3.07 in. |
| Length (Metric) | 78 mm |
| No. per Case | 1 |
| No. per Pack | 1 |
| Dimensions (L x W) | 78 x 610 m |
| Width (English) | 240 in. |
| Width (Metric) | 610 m |
| For Use With (Application) | Designed for applications ranging from PCR and real-time PCR to storage |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction