missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Microplate Sealing Film and Tapes

Product Code. 12762796 Shop All Axygen Scientific Products
Click to view available options
Material:
Aluminum
Litto or Ethylene or Vinyl
Polyester
Polyolefin
Rayon
Product Type:
AxySeal Sealing Film
Breathable Sealing Film
CyclerSeal Sealing Film
CyclerSeal Sealing Film Roll
Heat Seal Film
Sealing Film
Sealing Film Roll
UltraClear Sealing Film
Dimensions (L x W):
115 x 500 m
127.2 x 79.5 mm
133.3 x 79.2 mm
135.5 x 80 mm
146 x 79.2 mm
146 x 79.6 mm
153 x 82.3 mm
78 x 610 m
Not Available
Sterility:
Non-sterile
Sterile
Unit Size:
Each
Pack of 100
Pack of 1000
Pack of 50
Pack of 500
This item is not returnable. View return policy

Product Code. 12762796

Brand: Axygen™ PCRAS600

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Offer great economy for disposable ease.

For applications ranging from PCR and Real-Time PCR to ELISA and cell culture, Axygen films are an easy and cost-effective way to seal plates and enhance automation. Used to seal multi-well microplates. 

Aluminum

  • Provides exceptional seal on multi-well plates with varied temperatures and reagents
  • With compression mats, films and tapes seal during thermocycling
  • Ideal for light-sensitive samples and its uniformly applied adhesive provides optimum well sealing
  • Pierceable, PCR-AS-200

 

TRUSTED_SUSTAINABILITY

Specifications

Material Aluminum
Sterility Non-sterile
Length (English) 5.24 in.
Length (Metric) 133.3 mm
No. per Case 500/Cs.
No. per Pack 100/Pk.
Dimensions (L x W) 133.3 x 79.2 mm
Width (English) 3.11 in.
Width (Metric) 79.2 mm
For Use With (Application) Designed for applications ranging from PCR and real-time PCR to ELISA and cell culture, ideal for light-sensitive samples
Product Type Sealing Film
Thickness 50 μm
Temperature Range (Metric) -80°C to +104°C
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.