Learn More
Axygen™ Multi Well High Profile Reagent Reservoirs
Manufactured with virgin polypropylene for excellent chemical resistance
Brand: Axygen™ RES-MW4-HP
251.27 GBP valid until 2024-12-31
Use promo code "22009" to get your promotional price.
Description
Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.
- Durable and cost effective
- Various formats for multiple applications
Specifications
ANSI/SLAS, RNase/DNase-free, non-pyrogenic | |
50 mL | |
Non-sterile | |
25/Cs. | |
4 |
Clear | |
Polypropylene | |
Reagent Reservoir | |
12.5 mL | |
Single-use |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.