missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ PerkinElmer™ Compatible Tips
384 Well tips for use with PerkinElmer
Brand: Axygen PKF-384-R-S
Code : 68
Additional Details : Weight : 10.00000kg
Description
- DNAse-Free, RNAse-Free, Pyrogen-Safe
- 384-Tip format for MDT 384-30μL head
- Clear Polypropylene
- Sterile
- With Aerosol-Resistant Filters
- Packaging: 384 tips/rack, 10 racks/pk, 5 packs/case
Specifications
30 μL | |
Sterile | |
Not Autoclavable | |
Round | |
PerkinElmer |
Filtered | |
Clear | |
Rack | |
PerkinElmer | |
Polypropylene |