Learn More
Axygen™ PlateMax™ Semi-Automated Plate Sealer Accessory, Heat Sealing Films
GREENER_CHOICE
Designed for applications ranging from PCR and Real-Time PCR to ELISA and cell culture
Brand:  Axygen™ HS-200
 View more versions of this product
Product Code. 12627765
  • £527.45 / Pack of 500
Explore more special offers
Description
Description
Axygen PlateMax microplate heat sealing films are designed for applications ranging from PCR and Real-time PCR to storage. They can be used in a wide range of temperatures making them suitable for nearly any application.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Plate Sealing
Polyester, Aluminum
100/Pk.
PlateMax Plate Sealer
500/Cs.
Peelable Heat Sealing Film
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Axygen™ PlateMax™ Semi-Automated Plate Sealer Accessory, Heat Sealing Films > Compound Storage PCR; Polyester, Aluminum; -80 °C - 90 °C; 70 μm