Learn More
Axygen™ PlateMax™ Semi-Automated Plate Sealer Accessory, Heat Sealing Films
GREENER_CHOICE
Designed for applications ranging from PCR and Real-Time PCR to ELISA and cell culture
Brand:  Axygen™ HS-300
 View more versions of this product
Product Code. 12637765
  • £527.45 / Pack of 500
Explore more special offers
Description
Description
Axygen PlateMax microplate heat sealing films are designed for applications ranging from PCR and Real-time PCR to storage. They can be used in a wide range of temperatures making them suitable for nearly any application.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Plate Sealing
Polyester, Aluminum
100/Pk.
PlateMax Plate Sealer
500/Cs.
Pierceable Aluminum Heat Sealing Film
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Axygen™ PlateMax™ Semi-Automated Plate Sealer Accessory, Heat Sealing Films > Storage; Polyester, Aluminum; -80 °C - 120 °C; 20 μm