Learn More
Axygen™ PlateMax™ Semi-Automated Plate Sealer Accessory, Heat Sealing Films
GREENER_CHOICE
Designed for applications ranging from PCR and Real-Time PCR to ELISA and cell culture
Brand:  Axygen™ HS-150-QPCR
 View more versions of this product
Product Code. 12779618
  • £541.31 / Pack of 500
Explore more special offers
Description
Description
Axygen PlateMax microplate heat sealing films are designed for applications ranging from PCR and Real-time PCR to storage. They can be used in a wide range of temperatures making them suitable for nearly any application.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Plate Sealing
Polyester
100/Pk.
PlateMax Plate Sealer
500/Cs.
Peelable Heat Sealing Film for qPCR
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Axygen™ PlateMax™ Semi-Automated Plate Sealer Accessory, Heat Sealing Films > QPCR Fluorescence Colorimetric Measurements; Polyester; -80 °C - 80 °C; 100.5 μm