Learn More
Corning™ Axygen™ Polypropylene Automation Tip For PerkinElmer™ System
Brand: Corning™ RRP-200-CBK-R-S
Code : A0
Additional Details : Weight : 11.30000kg
Description
Order Info
dimQuantity
Specifications
200 μL | |
Black | |
Rack | |
PerkinElmer Automation System | |
Polypropylene |
Sterile | |
DNase, RNase Free | |
Automation, Round | |
Axygen |