missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Replacement Electrodes for 7cm Gel Box
Accessory for use with 7cm horizontal gel box
Brand: Axygen™ HGB7-NE
This item is not returnable.
View return policy
Description
Replacement electrodeSpecifications
Gel Box, 7 cm | |
Replacement Positive Electrode |