missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Black Hanging Format Automation Pipet Tips for Hamilton Microlab™ Star Line and Nimbus™
GREENER_CHOICEBIOPHARMA

Product Code. 12872283 Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
10 μL
50 μL
300 μL
1000 μL
Sterility:
Non-sterile
Sterile
Filtered:
Filtered
Non-filtered
Format:
Hanging Tip Rack
Stack Rack
Unit Size:
Pack of 11520
Pack of 1536
Pack of 2304
This item is not returnable. View return policy

Product Code. 12872283

Brand: Axygen™ HT300CBKHTRS

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Compatible with Hamilton MICROLAB™ STAR and Nimbus workstations

Axygen offers a comprehensive line of tips for Hamilton™ Microlab™ automation systems.

The tips are:

  • Manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality
  • RNase-/DNase-free and non-pyrogenic
  • Available in non-sterile, sterile, non-filtered and filtered
  • Conductive black polypropylene tips are available in 96-well hanging tip rack and nested tip rack and individual deck-seating rack format
  • Designed for Hamilton™ MICROLAB™ STAR, STARlet, STARplus and Nimbus™ workstations.
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 300 μL
Filtered Non-filtered
Sterility Sterile
Color Black
Autoclavable Not Autoclavable
Clean Claims DNase, RNase and Pyrogen free
For Use With (Application) The conductive, liquid level sensing tips are available in four sizes and specially designed for use with Hamilton™ liquid handling platforms, they enable precision pipetting across a wide range of applications
Format Hanging Tip Rack
Tip Style Automation, Round
Packaging 1 Rack/Pk, 24 Pks/Cs.
For Use With (Equipment) Hamilton MICROLAB STAR, Hamilton MICROLAB STARlet, Hamilton MICROLAB STARplus, Hamilton MICROLAB Nimbus
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.