Learn More
Axygen™ Black Hanging Format Automation Pipet Tips for Hamilton Microlab™ Star Line and Nimbus™
Compatible with Hamilton MICROLAB™ STAR and Nimbus workstations
Brand: Axygen™ HT-50-CBK-STK
This item is not returnable.
View return policy
Description
Axygen offers a comprehensive line of tips for Hamilton™ Microlab™ automation systems.
The tips are:
- Manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality
- RNase-/DNase-free and non-pyrogenic
- Available in non-sterile, sterile, non-filtered and filtered
- Conductive black polypropylene tips are available in 96-well hanging tip rack and nested tip rack and individual deck-seating rack format
- Designed for Hamilton™ MICROLAB™ STAR, STARlet, STARplus and Nimbus™ workstations.
Specifications
50 μL | |
Non-sterile | |
Not Autoclavable | |
The conductive, liquid level sensing tips are available in four sizes and specially designed for use with Hamilton™ liquid handling platforms, they enable precision pipetting across a wide range of applications | |
Automation, Round | |
120 Racks/Cs. | |
Polypropylene |
Non-filtered | |
Black | |
DNase, RNase and Pyrogen free | |
Stack Rack | |
4 Racks/Stack, 30 Stacks/Cs. | |
Hamilton MICROLAB STAR, Hamilton MICROLAB STARlet, Hamilton MICROLAB STARplus, Hamilton MICROLAB Nimbus |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Product Content Correction
Product Title
Axygen™ Black Hanging Format Automation Pipet Tips for Hamilton Microlab™ Star Line and Nimbus™ > 96-well tips, 50 μL, Non-filtered, Non-sterile, Stacked Tip Rack, Black
Spot an opportunity for improvement?Share a Content Correction