missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Rubber Dams for 7 cm Gel Box
Accessory for use with 7cm horizontal gel box
Brand: Axygen™ HGB7-CD
This item is not returnable.
View return policy
Description
- Set of 2
Specifications
Gel Box, 7 cm |