Learn More
Axygen™ Screw Cap Tubes without Caps: Self-Standing
Self-standing tubes
Brand:  Axygen™ ST-150-SS-X
 View more versions of this product
Product Code. 11354225
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Axygen microcentrifuge tubes feature a screw cap that provides a tight secure seal. Screw cap tubes are available in a variety of colors with a conical or self-standing bottom. Sterile options are also available.
  • Maximum G-force is 20,000 RCF.
  • RNase-/DNase-free
  • Nonpyrogenic
Tubes, with Cap, Sterile
All sterile screw cap microcentrifuge tubes are pre-assembled with non-looped caps with O-Rings.
Colored-cap versions are also available in sterile format.
Order Info
Caps sold separately.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Screw Cap
20,000 x g
Amber
500/Pk.
Non-sterile
Not Included
RNase, DNase, pyrogen-free
1.5 mL
Polypropylene
500 Tubes/Pk., 8 Packs/Cs.
No
Conical