missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Screw Caps with O-rings

Product Code. 12769368 Shop All Axygen Scientific Products
Click to view available options
Color:
Assorted
Blue
Brown
Clear
Gray
Green
Orange
Pink
Red
Violet
White
Yellow
Sterility:
Non-sterile
Sterile
Format:
Looped
Non-looped
Unit Size:
Pack of 4000
Pack of 500
This item is not returnable. View return policy

Product Code. 12769368

Brand: Axygen™ SCOBRS

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Select looped or non-looped format

Axygen microcentrifuge tubes feature a screw cap that provides a tight secure seal. Screw cap tubes are available in a variety of colors with a conical or self-standing bottom. Sterile options are also available.

  • Maximum G-force is 20,000 RCF.
  • RNase-/DNase-free
  • Nonpyrogenic


Tubes, with Cap, Sterile


All sterile screw cap microcentrifuge tubes are pre-assembled with non-looped caps with O-Rings.


Colored-cap versions are also available in sterile format.
TRUSTED_SUSTAINABILITY

Specifications

Includes O-ring
Material Polypropylene
Color Brown
Format Non-looped
Sterility Sterile
For Use With (Equipment) Both skirted and non-skirted variations of the 0.5 mL, 1.5 mL, and 2 mL microcentrifuge tubes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.