Learn More
Axygen™ Single-Channel Micropipette
Axygen™ Single-Channel Micropipette combines high levels of accuracy and precision with design innovations to make pipetting more comfortable during prolonged periods.
£229.00
Product Code | Brand | Accuracy | Precision | Volume (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Accuracy | Precision | Volume (Metric) | Price | Quantity & Availability | |||
10369589
|
Axygen™
AP-200 |
±1.2/±0.6% | ±0.6/±0.2% | 20 to 200 μL |
£229.00
Each |
In Stock
Log in to see stock. |
|||
10641121
|
Axygen™
AP-10 |
±4.0/±0.5% | ±2.8/±0.4μL | 0.5 to 10 μL |
£229.00
Each |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
Description
- Thumb-friendly soft spring pipetting system reduces fatigue
- Unique side EZIject button option reduces thumb movement
- Finger-hook facilitates relaxed grip
- Large push button is easy on the thumb
- Locking feature prevents accidental volume changes
Specifications
1 | |
Micropipette | |
1 Pack |