Learn More
Axygen™ Single Well Medium Profile Reagent Reservoirs
GREENER_CHOICE
Product Code. p-4910553 Shop All Axygen Scientific Products
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 25
Product Code. 12527877
Brand: Axygen™ RESSW384FX
Only null left
Add to basket
Add to basket
Manufactured with virgin polypropylene for excellent chemical resistance
Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.
  • Durable and cost effective
  • Various formats for multiple applications
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI/SLAS, RNase/DNase-free, non-pyrogenic
Color Clear
Capacity (Metric) 170 mL
Material Polypropylene
Sterility Non-sterile
Type Reagent Reservoir
No. per Case 25/Cs.
No. per Pack 25/Pk.
Packaging 25/Cs.
Channels 384
Disposable Single-use