missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Storage Microplates

Shop All Axygen Scientific Products


Click to view available options
Color:
Clear
Unit Size:
Pack of 50
Description
Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.
2 mL deep well plate ANSI/SLAS 1-2004: Microplates - Footprint Dimensions Ultra-flat surface ensures proper sealing with heat sealing film
Specifications
Specifications
| Certifications/Compliance | ANSI, SLAS 1-2004 |
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Autoclavable | Autoclavable |
| Color | Clear |
| Product Type | 96 Round Deep-Well Storage Microplate |
| Material | Polypropylene |
| Volume (Metric) Well | 1.8 mL |
| No. of Wells | 96 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction