missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Storage Microplates
Shop All Axygen Scientific Products

Click to view available options
Color:
Blue
Clear
Green
Sterility:
Non-sterile
Sterile
No. of Wells:
24
48
96
384
Volume (Metric) Well:
1.1 mL
1.8 mL
10,000 μL
1100 μL
125 μL
2000 μL
240 μL
5000 μL
550 μL
Unit Size:
Pack of 25
Pack of 30
Pack of 50
Description
Used for sample collection, in vitro growth chambers and long-term storage.
Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.
Excellent chemical resistance ANSI/SLAS 1-2004: Microplates - Footprint Dimensions Centrifugation to 4,000 RCF Diamond shaped wells allow complete sample recovery Black and white plates are available by special order Ultra-flat surface ensures proper sealing with heat sealing film
Specifications
Specifications
| Certifications/Compliance | ANSI, SLAS 1-2004 |
| Format | Standard |
| Lid | Without Lid |
| Array | 16 x 24 |
| Color | Clear |
| Material | Polypropylene |
| Volume (Metric) Well | 240 μL |
| No. of Wells | 384 |
| Sterility | Non-sterile |
| Well Shape | Square |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction